- Length:6"
- Material:Stainless Steel with Tungsten Carbide
- Curvature:Straight
- Cat. No.:76192-688
The Sklar® Tungsten Carbide Swedish needle holder is a ratchted finger ring instrument used in suturing.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The Tungsten Carbide Swedish needle holder features delicate jaws with pyramid-shaped teeth to ensure a firm grip and reduce needle rotation. The shanks of the Swedish needle holder are bent, similar to the Bozeman. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.