ContactUsLinkComponent
Sklarhone™ Stevens Tenotomy Scissors, OR Grade, Sklar®
Sklarhone™ Stevens Tenotomy Scissors, OR Grade, Sklar®
Catalog # 76192-946
Supplier:  Sklar
undefined
Sklarhone™ Stevens Tenotomy Scissors, OR Grade, Sklar®
Catalog # 76192-946
Supplier:  Sklar
Supplier Number:  23-1342
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Length:
    4.5"
  • Material:
    Stainless steel
  • Curvature:
    Curved
  • Cat. No.:
    76192-946
  • Blade type:
    Blunt/Blunt, Razor/Smooth
Specifications
About this item
Sklarhone™ Stevens Tenotomy Scissors are used for delicate dissection and cutting, commonly in ophthalmologic, neurological, and plastic surgery procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Tenotomy scissors have long handles and small, sharp blades with variable tip styles. The long handles allow the surgeon to have an adequate grip and control while dissecting or cutting in small or narrow areas. Sklarhone™ scissors feature one knife-like razor sharp edge and one standard scissor edge. The long handles allow the user to make the sharp incisions of a knife with the cutting ability of scissors. Catching skin and pulls are minimized by the use of Sklarhone™ premium scissors.