Specifications
- Length:3.5"
- Material:Stainless steel
- Curvature:Straight
- Cat. No.:76192-976
Specifications
About this item
Sklar® toothed Guilford-Wright suture wire cutting scissors are non-ratcheted, finger ring instruments used for cutting sutures in preparation for removal.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Blades are much shorter than the shanks. The overall length is shorter than the Smith.