ContactUsLinkComponent
Insect Rearing Chambers, Caron Products
Insect Rearing Chambers, Caron Products
Supplier:  Caron Products
   Sustainable Options Available
Certificates
undefined
Insect Rearing Chambers, Caron Products
Supplier:  Caron Products
   Sustainable Options Available
Specifications
  • Catalog No:
  • 76205-450
  • 76205-452
  • 76205-460
  • 76205-456
  • 76205-458
  • 76205-454
  • Environmentally Preferable:
  • Sustainable Packaging
  • Temperature Range:
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • Temperature Uniformity:
  • ±0.3 °C
  • ±0.3 °C
  • ±0.3 °C
  • ±0.3 °C
  • ±0.3 °C
  • ±0.3 °C
  • Temperature Sensor:
  • 3-Wire RTD
  • 3-Wire RTD
  • 3-Wire RTD
  • 3-Wire RTD
  • 3-Wire RTD
  • 3-Wire RTD
  • Temperature Control:
  • ±0.1 °C
  • ±0.1 °C
  • ±0.1 °C
  • ±0.1 °C
  • ±0.1 °C
  • ±0.1 °C
  • Humidity:
  • Optional
  • Optional
  • Optional
  • Optional
  • Optional
  • Optional
  • Refrigeration:
  • Electrostatic Coated Coil
  • Electrostatic Coated Coil
  • Electrostatic Coated Coil
  • Electrostatic Coated Coil
  • Electrostatic Coated Coil
  • Electrostatic Coated Coil
  • Environmental Certification:
  • CSA