ContactUsLinkComponent
Econo™ Spatula & Packer, Floor Grade, Sklar®
  76192-646
 :  Sklar
undefined
Econo™ Spatula & Packer, Floor Grade, Sklar®
  76192-646
 :  Sklar
 :  21-797
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Length:
    5.75"
  • Material:
    Stainless steel
  • Cat. No.:
    76192-646
The Econo™ double end spatula and packer is commonly used in podiatry.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
It features one smooth, rounded teardrop tip (spatula) and one blunt probe end (packer). The spatula end is used to separate the nail plate from the nail bed. The packer is used to place packing under or around the toe following a surgical procedure. Econo™ instruments are floor grade, disposable instruments that eliminate the need for cleaning and reprocessing. Because they are disposable, Econo™ instruments also reduce the likelihood of cross-contamination.