- Length:8"
- Material:Stainless Steel with Tungsten Carbide
- Curvature:Straight
- Cat. No.:76192-560
The Sklar® Cooley Tungsten Carbide Microvascular Needle Holder is a ratcheted, finger ring instrument designed to hold small sutures.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The Sklar® Cooley Tungsten Carbide Microvascular Needle Holder is commonly used in microsurgery, plastic surgery, and cardiovascular procedures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.