Specifications
- Length:5"
- Material:Stainless steel
- Curvature:Straight
- Cat. No.:76192-490
- Tip style:Serrated, Delicate
Specifications
About this item
Econo™ Semken Dressing Forceps are most commonly used for application and changing of dressings.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
These forceps feature narrow tips that may be beneficial for neurosurgery or other procedures performed in constricted areas. They may also be used for dissection of fine or single-layer tissue. Serrated tips provide a solid grip on both materials and tissue. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.