ContactUsLinkComponent
Econo™ Braun Episiotomy Scissors, Floor Grade, Sklar®
Econo™ Braun Episiotomy Scissors, Floor Grade, Sklar®
Catalog # 76192-464
Supplier:  Sklar
undefined
Econo™ Braun Episiotomy Scissors, Floor Grade, Sklar®
Catalog # 76192-464
Supplier:  Sklar
Supplier Number:  21-375
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Length:
    5.5"
  • Material:
    Stainless steel
  • Curvature:
    Angled
  • Cat. No.:
    76192-464
  • Blade type:
    Blunt/Blunt, Smooth
Specifications
About this item
Econo™ Braun Episiotomy Scissors, which may also known as Braun-Stadler Episiotomy Scissors, are used to perform episiotomies during labor and delivery.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ instruments are floor grade, disposable instruments sold in bulk quantities. These are non-ratcheted, finger ring scissors.