ContactUsLinkComponent
Biox Microtube Rack
Biox Microtube Rack
Catalog # 76208-062
Supplier:  TWD Scientific
undefined
Biox Microtube Rack
Catalog # 76208-062
Supplier:  TWD Scientific
Supplier Number:  BX90-8009
Specifications
  • No. of Holes:
    80
  • Color:
    Assorted
  • Description:
    Microtube Rack
  • Material:
    PP
  • Environmentally Preferable:
  • Cat. No.:
    76208-062
Specifications
About this item
Microcentrifuge Tube Racks are made of a high-strength polypropylene that is chemically resistant to alcohols and mild organic solvents. The molded 5×16, 80-Well array is designed for easy tube identification.
  • Polypropylene
  • Easy Mark writing area
  • Equipped with side handles
  • Assorted colors (red, blue, purple, green and yellow)
Microtube racks are designed for use with 1.5 or 2.0 mL microtubes. Racks feature a molded numeric grid for quick identification purposes.