Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
G418 disulphate solution

G418 disulphate solution

10 ml
Catalog # 10766-882
Supplier:   Mirus Bio
Supplier #:  MIR5920
G418 disulphate solution
Catalog # 10766-882
Supplier:  Mirus Bio
CAS Number:  108321-42-2
G418 disulphate solution

10 ml

CAS Number:  108321-42-2
Price
$125.84
Log in for availability
Each (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

G418 antibiotic solution is used for stable cell line generation, it ensures effective positive selection for cells expressing the neomycin resistance (neo) gene. Different cell types and cell culture conditions may require different concentrations of selection antibiotic.

  • Recommended working concentration range is 0.1 to 2.0 mg/mL in mammalian cells
  • Store at 4 °C
  • To be protected from light

Concentration: 50 mg/mL G418 Sulfate, sterile filtered in DI water.

Synonyms:  G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
  • CAS Number:108321-42-2
  • Formula:C₂₀H₄₄N₄O₁₈S₂
  • MDL Number:MFCD00058314
  • Molecular Weight:692.72 g/mol
Specifications

Specifications

  • Size:
    10 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.