Specifications
- Length:5.5"
- Material:Stainless Steel with Tungsten Carbide
- Curvature:Angled
- Cat. No.:76192-154
Specifications
About this item
Sklar Edge™ TC Fomon dorsal scissors are used in rhinoplasty procedures.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
The angled shanks reduce surgeon fatigue, and the curvature allows for manipulation of the soft tissue around the dorsum of the nose. Available individually, or as a component of Sklar's major rhinoplasty set. Sklar Edge™ Tungsten Carbide Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades, which maximizes control and precision. Sklar Edge™ scissors can be identified by two gold coated finger rings.