ContactUsLinkComponent
KWIK-STIK Plus, Microbiologics
KWIK-STIK Plus, Microbiologics
Catalog # 76199-860
CAS Number:  
undefined
KWIK-STIK Plus, Microbiologics
Catalog # 76199-860
Supplier Number:  01209X
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Shigella boydii (2) derived from ATCC® 8700™*
  • Cat. No.:
    76199-860
Specifications
About this item
Combines microorganisms that are just two passages from their reference strains, with our signature KWIK-STIK format.
  • Over 900 strains available
  • All-in-one design reduces the risk of contamination
  • Strains are less than or equal to four passages from reference culture
  • Ready-to-use format saves time and money
  • Refrigerated storage is easy and economical
  • Online Certificate of Analysis provides detailed strain information
  • Traceability to reference cultures ensures authenticity
  • Technical Support experts available for guidance
Some laboratory applications require strains that are a fewer passages from the original reference culture. For those special instances, we offer KWIK-STIK Plus, which combines microorganisms that are just two passages from their reference strains, with our signature KWIK-STIK format. Each device features a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab; so everything you need for growing control cultures is included in one handy device.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.