ViroMag™ CRISPR, Transduction Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
ViroMag CRISPR reagent is the only magnetic viral transduction enhancer for CRISPR/Cas viruses (adenovirus, lentivirus, retrovirus) that transduces cells for gene editing applications.
- Highly increase transduction efficiency of viral CRISPR/Cas9 system
- Enable genome editing even in primary, hard to transduce and non-permissive cells
- Accelerate the transduction process
- Concentrate the viral dose onto the cells
- Synchronize cells absorption/infection without modifications of the viruses
Based on the Magnetofection™ technology, this reagent allows concentrating the complete applied dose of viral particles onto cells within minutes, inducing a significant improvement of virus infectivity with extremely low vector doses. Due to its specific properties, ViroMag CRISPR reagent is also ideal to infect non permissive cells. This reagent demonstrates an exceptionally high efficiency to promote, control and assist viral transductions so that no molecular biology processes or biochemical modifications are required.
Frequently Bought Together