Order Entry
Puerto Rico
ContactUsLinkComponent
Streptococcus pyogenes Cas9 Nuclease, OZ Biosciences
Streptococcus pyogenes Cas9 Nuclease, OZ Biosciences
Supplier:  OZ Biosciences
Certificates
undefined
Streptococcus pyogenes Cas9 Nuclease, OZ Biosciences
Supplier:  OZ Biosciences

Specifications

  • Species:
    Streptococcus pyogenes
  • Storage Conditions:
    −20 °C
  • Form:
    Lyophilized powder
  • Enzyme Name:
    Cas9 Nuclease

Specifications

Related Information

Frequently Bought Together