Order Entry
Puerto Rico
ContactUsLinkComponent
 
FluoProdige Assay Kit, OZ Biosciences
  103537-344
 :  
undefined
FluoProdige Assay Kit, OZ Biosciences
  103537-344
 :  FPR0200
 :  

 

  • Cat. No.:
    103537-344
  • No. of assays/kit:
    2000
  • Includes:
    FluoProdige fluorescent Dye (10 mL), Dilution Buffer (100 mL), BSA (500 µg) lyophilized

 

 

The FluoProdige Protein Quantification Assay Kit presents a complete fluorometric assay for protein and peptide quantification. FluoProdige facilitates and improves protein quantification by providing sensitivity and convenience when compared to classic colorimetric measurements such as Lowry or BCA.

  • Accurate and highly sensitive: Detect as little as 40 ng/mL - Large linear detection range over 3-orders of magnitude
  • Simple and Fast: Protocol is 15 min - Pic of signals within 30 min - Signal duration of up to 4 hours
  • Safe and Biodegradable: Natural product, non-toxic, heavy metal free
  • Compatible with many buffers: Robust to interfering compounds such as DNA, Detergents, DTT and Urea

This assay kit uses a stable analogue to epicocconone molecule that reversibly binds to lysine, arginine and histidine residues in proteins and peptides to yield an intense red-fluorescent product. The fluorescent signal (~518/605) is directly proportional to protein amount among a wide range of protein concentration, rendering this kit highly sensitive. When bound to proteins, excitation and emission signals are 518/605 nm respectively.

 : Shipped at room temperature.
 : Upon receipt, store all reagent tubes at 4 °C (up to 6 months). For long term storage, store the dye at –20 °C.