Order Entry
Puerto Rico
ContactUsLinkComponent
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) substrate ≥99%, Molecular Biology Grade
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) substrate ≥99%, Molecular Biology Grade
Catalog # 103258-198
Supplier:  OZ Biosciences
CAS Number:  
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) substrate ≥99%, Molecular Biology Grade
Catalog # 103258-198
Supplier:  OZ Biosciences
Supplier Number:  XG31000
CAS Number:  

Specifications

  • Size:
    1 g
  • Cat. No.:
    103258-198
  • Supplier no.:
    XG31000

Specifications

About this item

 :
 :408.63 g/mol
Storage Temperature:Freezer
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666