Sterilization Indicating Labels, Brady
This B-7425-AC material is a steam sterilization indicating polypropylene label with a preprinted header that displays 'STERILIZED' when exposed to autoclave conditions.
- Adhesive type: acrylic
- Suggested ribbon series: R4300, R6200, R6400
- Thickness: 0.004"
- Chemical-resistant, cold-resistant, high temperature-resistant, and low temperature-resistant
Ideal for use in sample tracking, inventory management, general tracking, and identification of lab materials that require sterilization. An economical solution for identifying vials, tubes, racks, and flasks. Labels adhere to stainless steel, glass, and polypropylene, and will withstand temperatures from –320.8 to 249.8 °F (–196 to 121 °C ).