ContactUsLinkComponent
NOVA-TECH™ P Nonwoven Polypropylene Cleanroom Wipes, High-Tech Conversions
NOVA-TECH™ P Nonwoven Polypropylene Cleanroom Wipes, High-Tech Conversions
Catalog # 76201-720
undefined
NOVA-TECH™ P Nonwoven Polypropylene Cleanroom Wipes, High-Tech Conversions
Catalog # 76201-720
Supplier Number:  NTP-1111
Specifications
  • Description:
    NOVA-TECH™ P Wipes
  • Material:
    PP
  • Cat. No.:
    76201-720
  • Packaging:
    500 Wipes/Bag, 6 Bags/Case
  • W×L:
    27.9 x 27.9 cm (11 x 11")
Specifications
About this item
NOVA-TECH™ P Wipes are made from 100% polypropylene and provide an extremely clean and absorbent solution to cleanroom needs.
  • 100% polypropylene
  • Exremely clean
  • High absorbency
  • Cleanroom packaging
These wipes can be used in a wide range of industries and applications, such as life sciences, pharmaceutical labs, aerospace industries, and much more. The large surface area allows for use on larger spills or tasks within any working environment that smaller wipes are not able to handle.