Order Entry
Puerto Rico
ContactUsLinkComponent
ID-Vit® Folic Acid Vitamin B9 Microbiological Test Kit, Alpco
ID-Vit® Folic Acid Vitamin B9 Microbiological Test Kit, Alpco
Catalog # 101721-416
Supplier:  Alpco Diagnostics
CAS Number:  
ID-Vit® Folic Acid Vitamin B9 Microbiological Test Kit, Alpco
Catalog # 101721-416
Supplier:  Alpco Diagnostics
Supplier Number:  30-KIF005
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    ID-Vit® Folic Acid Vitamin B9 Microbiological Test Kit
  • Size:
    100 µl
  • Cat. No.:
    101721-416
  • Sensitivity:
    4 µg/l
  • No. of wells:
    96 wells
  • Sample type:
    Serum
  • Application:
    ELISA
  • Range:
    4 – 40 µg/l
  • Supplier no.:
    30-KIF005

Specifications

About this item

ID-Vit® Folic acid is a microtiter plate test kit based on a microbiological method which measures the total folic acid content in serum.

The test kit contains the standard and all reagents required to perform the test. It is sufficient for 96 determinations including standard curves.

Delivery: Kit includes Lactobacillus rhamnosus (precoated microtiter plate), 4 x 5 mL sample treatment solution, 4 x 30 mL water, 4 x folic acid assay medium, 4 x folic acid standard, lyoph., 4 x adhesive cover foil, replacement holder for microtiter strips, 4 x 1.5 mL folic acid medium treatment buffer, 4 x folic acid control 1, lyoph. and 4 x folic acid control 2, lyoph.
Caution: For use in human and veterinary medicine and in research. For in vitro diagnostic use only.