Order Entry
Puerto Rico
ContactUsLinkComponent
pGEM-3Z Vector, 20 µg, Promega
pGEM-3Z Vector, 20 µg, Promega
Catalog # PAP2151
CAS Number:  
undefined
pGEM-3Z Vector, 20 µg, Promega
Catalog # PAP2151
Supplier Number:  P2151
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAP2151
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    P2151

Specifications

About this item

Intended for use as a standard cloning vector or for high-fidelity efficient synthesis of RNA in vitro.

  • Standard Cloning Vector that can Efficiently Synthesize RNA In vitro
  • Blue/white screening to easily identify recombinant clones
  • Versatile vector used for standard cloning and in vitro transcription from SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • Multiple cloning site provides a convenient selection of restriction sites for cloning

The pGEM-3Z Vector is intended for use as a standard cloning vector, as well as for the highly efficient synthesis of RNA in vitro. The vector carries the lacZ alpha-peptide and the multiple cloning region arrangement from pUC18 allowing selection of recombinants by blue/white screening. In addition, the vector contains both the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region. The pGEM-3Z and pGEM-4Z Vectors are essentially identical except for the orientation of the SP6 and T7 promoters.