Order Entry
Puerto Rico
ContactUsLinkComponent
pNLCoI1[luc2-P2A-NlucP/Hygro] Vector, 20 µg, Promega
pNLCoI1[luc2-P2A-NlucP/Hygro] Vector, 20 µg, Promega
Catalog # PAN1461
CAS Number:  
undefined
pNLCoI1[luc2-P2A-NlucP/Hygro] Vector, 20 µg, Promega
Catalog # PAN1461
Supplier Number:  N1461
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAN1461
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    N1461

Specifications

About this item

The pNLCoI Vectors comprise a second-generation coincidence reporter vector system that allow expression of both firefly luciferase (luc2) and NanoLuc Luciferase fused to a PEST destabilization domain (NlucP) from the same mRNA transcript.

  • Vectors That Express Both Firefly and NanoLuc(R) Luciferases from the Same Transcript
  • Firefly and NanoLuc(R) luciferase have dissimilar compound interference profiles for better identifying more false-positives versus individually
  • Using two different transcriptional reporters reduces false hit rates and increases the identification of true biological hits
  • luc2 and NlucP provide a bright reporter combination compatible with low-copy-number and plate scale-up, and provide greater signal-to-background ratios

Luciferase-based reporter-gene assays remain a useful and powerful method of high-throughput compound screening. However, false hits that result from direct interaction of compounds with the luciferase reporter can result in unnecessary follow-up efforts. The pNLCoI Vectors comprise a second-generation coincidence reporter vector system that allow expression of both firefly luciferase (luc2) and NanoLuc Luciferase fused to a PEST destabilization domain (NlucP) from the same mRNA transcript. The stoichiometric expression of both luciferases is achieved by use of the P2A sequence from porcine teschovirus-1, which promotes a ribosomal skip and expression of the two unfused enzymes with distinct compound interaction profiles. When used in high-throughput compound screening, false hits caused by direct interaction with one or the other luciferases can be distinguished from true hits that show a similar response for both, reducing workload associated with follow-up screens. The pNLCoI Vectors are designed for use with the Nano-Glo Dual-Luciferase Reporter (NanoDLR) Assay System, which allows sequential detection of firefly and NanoLuc Luciferase in activity in the same sample. Both reagents provide stable glow-type luminescence signals with half-lives of approximately two hours allowing batch processing of samples and amenable to assays or screens in 96-, 384- or 1,536-well plate formats. Potent inhibition of firefly luciferase coupled with the high-intensity luminescence of NanoLuc luciferase maximizes sensitivity for detection of both reporters.