Order Entry
Puerto Rico
ContactUsLinkComponent
pNL1.1.PGK[Nluc/PGK] Vector, 20 µg, Promega
pNL1.1.PGK[Nluc/PGK] Vector, 20 µg, Promega
Catalog # PAN1441
CAS Number:  
undefined
pNL1.1.PGK[Nluc/PGK] Vector, 20 µg, Promega
Catalog # PAN1441
Supplier Number:  N1441
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAN1441
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    N1441

Specifications

About this item

The promoter-driven NanoLuc (Nluc) control vectors can be used to co-transfect with experimental firefly luciferase vectors when using the Nano-Glo Dual-Luciferase Reporter (NanoDLR) Assay System.

  • Vectors Containing NanoLuc(R) Luciferase Expressed from Mammalian Promoters
  • Multiple promoter options (CMV, TK and PGK) are available to obtain appropriate levels of the control reporter in your experimental system
  • Co-transfect with experimental firefly luciferase vectors as normalization control
  • Measure signal using the Nano-Glo(R) Dual-Luciferase(R) Reporter (NanoDLR™) Assay System

The promoter-driven NanoLuc (Nluc) control vectors can be used to co-transfect with experimental firefly luciferase vectors when using the Nano-Glo Dual-Luciferase Reporter (NanoDLR) Assay System. NanoLuc luciferase is a small (19.1kDa), stable reporter enzyme that can be up to 100-fold more sensitive than the flash-type Renilla signal in the DLR Assay and more than 3,000-fold more sensitive than the Renilla signal in the Dual-Glo Assay. The increased brightness of the NanoLuc Luciferase means you use less control DNA, minimizing assay artifacts and providing a stable control signal for normalization of the experimental Fluc reporter. The vectors are engineered with minimal consensus transcription factor-binding sites to reduce anomalous expression.