Order Entry
Puerto Rico
ContactUsLinkComponent
pNLF1-N [CMV/Hygro] Vector, 20 µg, Promega
pNLF1-N [CMV/Hygro] Vector, 20 µg, Promega
Catalog # PAN1351
CAS Number:  
undefined
pNLF1-N [CMV/Hygro] Vector, 20 µg, Promega
Catalog # PAN1351
Supplier Number:  N1351
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAN1351
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    N1351

Specifications

About this item

The three pNLF1 Vectors create N- or C-terminal fusions of NanoLuc luciferase and your protein of interest using a multiple cloning site (MCS). Insert your protein on the N or C terminus of NanoLuc luciferase or the N terminus of secreted NanoLuc enzyme.

  • Generate N- or C-Terminal Fusions with NanoLuc(R) Enzyme or N-Terminal Secreted NanoLuc(R) Luciferase
  • Use traditional cloning to insert protein of interest into the multiple cloning site (MCS)
  • Bright NanoLuc(R) reporter enables endogenous expression levels of fusion proteins for biologically relevant results
  • Choose from transient transfection or select for stable expression using hygromycin

The small size (19.1kDa) and extreme brightness (about 100-fold brighter than either firefly [Photinus pyralis] or Renilla reniformis) of NanoLuc luciferase (Nluc) make it an ideal protein fusion partner. NanoLuc fusion proteins can be used in a variety of applications including: reporters of protein stability, probes for bioluminescent cell imaging (BLI) or as the donor signal in bioluminescent resonance energy transfer (BRET) applications for protein:protein or protein:small-molecule interaction studies. The NanoLuc protein fusion vectors enable simple generation of N- or C-terminal fusions of NanoLuc luciferase with your protein of interest. The pNLF Vector series uses using traditional cloning with a multiple cloning site (MCS) to generate N- or C-terminal fusions to the full-length Nluc protein with the pNLF1-N or pNLF1-C [CMV/Hygro] Vectors or attach secreted Nluc to the N-terminus of the protein of interest with pNLF1-secN [CMV/Hygro] Vector. Also make NanoLuc protein fusions with the pF Vector series: Generate N- or C-terminal Nluc fusion proteins using the Flexi Vector Cloning System—a directional cloning method based on two rare-cutting restriction enzymes, SgfI and PmeI, that provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence.