Order Entry
Puerto Rico
ContactUsLinkComponent
pFC32K Nluc CMV-neo Flexi Vector, 20 µg, Promega
pFC32K Nluc CMV-neo Flexi Vector, 20 µg, Promega
Catalog # PAN1341
CAS Number:  
undefined
pFC32K Nluc CMV-neo Flexi Vector, 20 µg, Promega
Catalog # PAN1341
Supplier Number:  N1341
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAN1341
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    N1341

Specifications

About this item

The pFN31 and pFC32 Vectors generate N- or C-terminal fusions of NanoLuc luciferase with your protein of interest. Use pFN31A/K Nluc CMV-Hygro Flexi Vectors for N-terminal fusions and pFC32A/K Nluc CMV-Hygro Flexi Vectors for C-terminal fusions.

  • Generate N- or C-Terminal Fusions to NanoLuc(R) Luciferase Reporter
  • Insert protein of interest using the Flexi(R) Vector System
  • Select options for N- or C-terminal NanoLuc(R) fusions and antibiotic resistance (ampicillin or kanamycin)
  • Bright NanoLuc(R) reporter enables endogenous expression levels of fusion proteins for biologically relevant results

The small size (19.1kDa) and extreme brightness (about 100-fold brighter than either firefly [Photinus pyralis] or Renilla reniformis) of NanoLuc luciferase (Nluc) make it an ideal protein fusion partner. NanoLuc fusion proteins can be used in a variety of applications including: reporters of protein stability, probes for bioluminescent cell imaging (BLI) or as the donor signal in bioluminescent resonance energy transfer (BRET) applications for protein:protein or protein:small-molecule interaction studies. The NanoLuc protein fusion vectors enable simple generation of N- or C-terminal fusions of NanoLuc luciferase with your protein of interest and are available. The pF Vector series generates N- or C-terminal Nluc fusion proteins using the Flexi Vector Cloning System—a directional cloning method based on two rare-cutting restriction enzymes, SgfI and PmeI, that provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. Select the pFN31A and pFN31K Nluc CMV-Hygro Flexi Vectors for N-terminal protein fusions and pFC32A and pFC32K Nluc CMV-Hygro Flexi Vectors for C-terminal protein fusions. Also make NanoLuc protein fusions with the pNLF Vector series: Generate N- or C-terminal fusions to the full-length Nluc protein or attach secreted Nluc to the N-terminus of the protein of interest using traditional cloning with a multiple cloning site (MCS).