Order Entry
United States
ContactUsLinkComponent
pNL3.2[NlucP/minP] Vector, 20 µg, Promega
pNL3.2[NlucP/minP] Vector, 20 µg, Promega
Catalog # PAN1041
CAS Number:  
undefined
pNL3.2[NlucP/minP] Vector, 20 µg, Promega
Catalog # PAN1041
Supplier Number:  N1041
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    20 μg
  • Environmentally Preferable:
  • Cat. No.:
    PAN1041
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    N1041

Specifications

About this item

NanoLuc (Nluc) luciferase is a small enzyme (19.1kDa) engineered for optimal performance as a luminescent reporter. The pNL3.1[Nluc/minP] and pNL3.2[NlucP/minP] Vectors offer a minimal promoter for cloning response elements of interest.

  • Clone Response Elements of Interest to Drive Expression of NanoLuc(R) Luciferase
  • Nluc genes are codon optimized for mammalian cells
  • Many potential regulatory elements or other undesirable features have been removed (such as common restriction enzyme sites)
  • Vectors have a pGL4-based backbone for easy sequence transfer from existing plasmids

NanoLuc (Nluc) luciferase is a small enzyme (19.1kDa) engineered for optimal performance as a luminescent reporter. The enzyme is about 100-fold brighter than either firefly (Photinus pyralis) or Renilla reniformis luciferase using a novel substrate, furimazine, to produce high intensity, glow-type luminescence. The luminescent reaction is ATP-independent and designed to suppress background luminescence for maximal assay sensitivity. The pNL3.1[Nluc/minP] and pNL3.2[NlucP/minP] Vectors offer a minimal promoter for cloning response elements of interest. For use as a genetic reporter, multiple forms of NanoLuc luciferase have been configured to meet differing experimental objectives. Unfused Nluc offers maximal light output and sensitivity, NanoLuc-PEST (NlucP) closely couples protein expression to changes in transcriptional activity and increased signal-to background ratios, and NanoLuc luciferase fused to an N-terminal secretion signal (secNluc) is suitable when a secreted reporter is preferred. Luminescence is linearly proportional to the amount of NanoLuc protein over a 1,000,000-fold concentration range, with a signal half-life >/=2 hours when detected with Nano-Glo Luciferase Assay Reagent. NanoLuc luciferase possesses a number of physical properties that make it an excellent reporter protein: 1) very small, monomeric enzyme (171 amino acids; 513bp); 2) high thermal stability (Tm = 60°C); 3) active over a broad pH range (pH 6-8); 4) no post-translational modifications or disulfide bonds; 5) uniform distribution in cells; 6) emission spectrum well suited for bioluminescence resonance energy transfer (BRET; lambdamax = 465nM). NanoLuc luciferase is made available in a variety of plasmids designed for use in reporter gene assays of transcriptional control and with each of the NanoLuc forms (unfused Nluc, PEST destabilized NlucP, and secreted secNluc). The different pNL variations are designed for the following: 1) pNL1: cloning of a known or putative promoter region; 2) pNL2: cloning of a known or putative promoter region and establishment of a stable cell line through Hygromycin selection; 3) pNL3: cloning of a binding site or response element not in need of a basic promoter (such as are present in the pNL3.2.NF-kB-RE vector); 4) Control plasmids for the unfused, PEST-destabilized and secreted Nluc forms also are available. The pNL vector series uses a pGL4-based backbone for easy sequence transfer from existing plasmids. This backbone design also reduces anomalous results by removing many transcription factor binding sites and other potential regulatory elements. The Nluc gene variations are codon optimized and have had many potential regulatory elements or other undesirable features removed (such as common restriction enzyme sites).