Order Entry
Puerto Rico
ContactUsLinkComponent
pF25K ICE T7 Flexi Vector, 20 µg, Promega
pF25K ICE T7 Flexi Vector, 20 µg, Promega
Catalog # PAL1081
CAS Number:  
undefined
pF25K ICE T7 Flexi Vector, 20 µg, Promega
Catalog # PAL1081
Supplier Number:  L1081
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAL1081
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    L1081

Specifications

About this item

Designed for use with Insect Cell Extracts and contain untranslated region (UTR) sequences at the 5´ and 3´ ends of the gene coding region to enhance translation efficiency.

  • Protein Expression Vectors Used With Insect Cell Extracts for In Vitro Translation
  • Efficient transfer of Flexi(R) vector inserts for direct use of recombinant clones, minimizing time wasted screening background colonies
  • Adaptable to high-throughput formats for large screening projects
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or mammalian protein expression) as needed

The Flexi Vector System is a simple, yet powerful, directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. The vectors are designed with special sequences for maximal cell-free protein expression in a specific system. The pF25A/K ICE Vectors were designed for use with Insect Cell Extracts and contain untranslated region (UTR) sequences at the 5´ and 3´ ends of the gene coding region to enhance translation efficiency. Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert.