Order Entry
Puerto Rico
ContactUsLinkComponent
pFN28K HaloTag CMV-neo Flexi Vector, 20 µg, Promega
pFN28K HaloTag CMV-neo Flexi Vector, 20 µg, Promega
Catalog # PAG8451
CAS Number:  
undefined
pFN28K HaloTag CMV-neo Flexi Vector, 20 µg, Promega
Catalog # PAG8451
Supplier Number:  G8451
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAG8451
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    G8451

Specifications

About this item

Vectors designed for expression of N-terminal-tagged HaloTag fusion proteins in mammalian cells. Once expressed, use the HaloTag fusion protein for cell imaging of protein localization or trafficking in conjunction with the fluorescent HaloTag Ligands.

  • Append N-Terminal Tag for High-Level Protein Expression in Mammalian Cells
  • Enables constitutive protein expression in mammalian cells
  • Includes CMV immediate-early enhancer/promoter
  • Suited to stable or transient gene expression

These vectors are designed for expression of N-terminal-tagged HaloTag fusion proteins in mammalian cells. Once expressed, the HaloTag fusion protein may be used for cell imaging of protein localization or trafficking in conjunction with the fluorescent HaloTag Ligands. In addition, the HaloTag fusion protein can be purified or pulled down as a complex with its protein partners. We offer two types of HaloTag fusion vectors to accommodate your cloning preferences: pHT Vector Series: Simple Multiple Cloning Site (MCS) plasmids for traditional cloning. pF Vector Series: Flexi Vector Cloning System---a directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert. Find your gene, precloned, and experimentally validated at: www.promega.com/products/pm/halotag-technology/kazusa-collection