ContactUsLinkComponent
pGL4.54[luc2/TK] Vector, 20 µg, Promega
  PAE5061
 :  
undefined
pGL4.54[luc2/TK] Vector, 20 µg, Promega
  PAE5061
 :  E5061
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Size:
    20 μg
  • Cat. No.:
    PAE5061
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    E5061
pGL4.13[luc2/SV40] ,pGL4.53[luc2/PGK] and pGL4.54[luc2/TK] Vectors are used as controls in dual-luciferase assays where Renilla or NanoLuc luciferase is the experimental reporter. Select the appropriate promoter or test all 3 in your experimental system.
  • Vectors That Are Co-transfected into Cells as Controls in Dual-Reporter Assays
  • Co-transfect into cells where Renilla or NanoLuc(R) luciferase is the experimental reporter gene
  • SV40, PGK and TK promoter options offer different expression levels suitable for most dual-reporter assays
The Promoter-driven firefly control vectors are commonly co-transfected with experimental Renilla or NanoLuc luciferase vectors for use in the Dual-Luciferase or Dual-Glo Reporter Assay Systems. The control firefly vectors should give an almost invariant level of activity, while the experimental Renilla or NanoLuc vector varies with treatment. The pGL4.13[luc2/SV40] ,pGL4.53[luc2/PGK] and pGL4.54[luc2/TK] Vectors offer three different promoter options for your dual-reporter assay.