qPCR Consumables, Agilent Technologies
Supplier: Agilent Technologies
Certificates
About this item
These QPCR Plastics and accessories have been validated for optimal performance with Mx3005P, Mx3000P and the AriaMx real time PCR Systems.
Plates and tubes fit most PCR instruments. Racks hold standard PCR and microcentrifuge tubes.
The StrataCooler LP benchtop cooler provides the most protection for enzymes and the cooler will maintain a temperature measuring as low as –15 °C even after 2 hours.
StrataCooler Cryo preservation modules enable highly controlled freezing (0.4 to 0.6 °C/minute) of mammalian and insect cells to significantly increase survival rates (up to 90%).
96-Well Semi-skirted Polypropylene PCR plate validated for optimal performance and compatible with the optical Strip Caps.
Caution: For research use only. Not for use in diagnostic procedures.