Order Entry
Puerto Rico
ContactUsLinkComponent
pSV-beta-Galactosidase Control Vector, 20 µg, Promega
pSV-beta-Galactosidase Control Vector, 20 µg, Promega
Catalog # PAE1081
CAS Number:  
undefined
pSV-beta-Galactosidase Control Vector, 20 µg, Promega
Catalog # PAE1081
Supplier Number:  E1081
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAE1081
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    E1081

Specifications

About this item

A positive control vector for monitoring transfection efficiencies of mammalian cells. The SV40 early promoter and enhancer drive transcription of the lacZ gene, which encodes the beta-galactosidase enzyme.

  • Monitors Transfection Efficiencies of Mammalian Cells
  • Co-transfect into cells as the control reporter in a dual-reporter assay
  • Assay in cell extracts using spectrophotometric, fluorescent or chemiluminescent methods
  • Can be used for in situ histochemical analysis using X-gal substrate

The pSV-Beta-Galactosidase Control Vector is a positive control vector for monitoring transfection efficiencies of mammalian cells. The SV40 early promoter and enhancer drive transcription of the lacZ gene, which encodes the beta-galactosidase enzyme. The pSV-Beta-Galactosidase Control Vector can be transfected individually or co-transfected with your DNA of interest. Beta-galactosidase is an excellent reporter enzyme that can be assayed quickly and directly in cell extracts using spectrophotometric, fluorescent or chemiluminescent assays. This reporter enzyme also is widely used for in situ histochemical analysis using the substrate X-Gal. The pSV-Beta-Galactosidase Control Vector can be co-transfected with your DNA of interest. For example, co-transfection with firefly luciferase gene vectors (pGL3 Vectors) provide cell extracts that can be assayed for both luciferase and beta-galactosidase activities. In this manner, the pSV-Beta-Galactosidase Vector acts as an internal control for transient expression assays. A negative control extract, prepared from mock-transfected cells, should also be assayed for the presence of endogenous beta-galactosidase activity in cultured cells. In addition, co-transfection with chloramphenicol acetyltransferase reporter gene vectors (pCAT3 Vectors) permits assaying for both CAT and beta-galactosidase activities. The pSV-Beta-Galactosidase Vector is a modification of pRSV-beta-Gal with SV40 and pUC18 sequences substituted for RSV and pBR322 sequences. The pSV-Beta-Galactosidase Vector will express beta-galactosidase in E. coli due to the presence of the E. coli gpt promoter located upstream of the lacZ gene. Colonies of E. coli containing the pSV-Beta-Galactosidase Vector will appear blue when plated on media containing X-Gal.