Order Entry
Puerto Rico
ContactUsLinkComponent
 
Rat Recombinant IL-3 B (from E. coli)
 
undefined
Rat Recombinant IL-3 B (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity Assay: NFS-60 cell proliferation, ED₅₀ ≤10 ng/ml, Bioactivity ≥1.0×10⁴ units/mg
  • Gene ID:
    P04823
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    MCGF|P-cell stimulating factor|HCGF|Multi-CSF
  • Protein/Peptide Name:
    IL-3 B
  • Purity:
    ≥95%
  • Molecular Weight:
    16.3 kDa
  • Sequence:
    MISDRGSDAH HLLRTLDCRT IALEILVKLP YPQVSGLNNS DDKANLRNST LRRVNLDEFL KSQEEFDSQD TTDIKSKLQK LKCCIPAAAS DSVLPGVYNK DLDDFKKKLR FYVIHLKDLQ PVSVSRPPQP TSSSDNFRPM TVEC
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay