Mouse Recombinant IGF-II (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Insulin-like growth factor II (IGF-II) is an important fetal growth hormone made by theca cells during gestation. IGF-II engages the IGF-I receptor (IGF1R) to mediate embryonic growth. IGF-II also binds the sink IGF-II receptor (IGF2R) leading to IGF-II degradation.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:−20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: FDC-P1 cell proliferation, ED50 ≤50 ng/ml, bioactivity ≥2.0×10⁴ units/mg
- Gene ID:P09535
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:Somatomedin A
- Protein/Peptide Name:IGF-II
- Purity:≥95%
- Molecular Weight:7.4 kDa
- Sequence:AYGPGETLCG GELVDTLQFV CSDRGFYFSR PSSRANRRSR GIVEECCFRS CDLALLETYC ATPAKSE
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Frequently Bought Together