StrataPrep PCR Purification Kits, Agilent Technologies
: Avantor
The StrataPrep PCR purification kits provide a rapid method to simultaneously purify PCR products from PCR primers, unincorporated nucleotides, buffer components, and enzymes.
- Accommodates purification of PCR products from 100 bp to long PCR amplicons
- Highly purified DNA, ready for use
- No hazardous organic extraction or ethanol precipitation
- Ideal purification for high throughput sequencing and microarrays
The StrataPrep PCR purification kits removes Contaminants Such as Primers and Non-Specific Amplification Products From Reactions With Yields of Up To 10 µg. Purified DNA Exhibits Twice The Cloning Efficiency of Competitor Products.
: For research use only. Not for use in diagnostic procedures.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Amplified/modified DNA
- Sample type:PCR and other enzymatic reactions
- Target molecule:DNA
- Description:StrataPrep PCR Purification Kit
Frequently Bought Together