Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Brilliant Probe Multiplex MM, Agilent Technologies

Brilliant Probe Multiplex MM, Agilent Technologies

Brilliant Multiplex QPCR Master Mix, 200× 25 µl
Catalog # ST600553
Supplier #:  600553
Brilliant Probe Multiplex MM, Agilent Technologies
Catalog # ST600553
CAS Number:  
Brilliant Probe Multiplex MM, Agilent Technologies
Brilliant Probe Multiplex MM, Agilent Technologies

Brilliant Multiplex QPCR Master Mix, 200× 25 µl

CAS Number:  
Price
$477.31
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

The brilliant multiplex QPCR master mix allows you to amplify up to four targets in a single real-time PCR Reaction.

  • Maximizes analysis of limited or rare samples
  • Allows detection of multiple targets plus an internal control
  • More economical per sample than singleplex, while saving time and increasing throughput

It provides sufficient reaction components to accurately quantify both low-and-high-abundance targets in the same tube.

The sensitivity remains equivalent to that seen in singleplex.

Specifications

Specifications

  • Description:
    Brilliant Multiplex QPCR Master Mix
  • No. of reactions:
    200× 25 µl
  • Multiplexing:
    Yes
  • Cat. No.:
    ST600553
  • Application:
    qPCR probe

Specifications