Human Recombinant GRO-beta (from E. coli)
Produced in E. coli. Contains 73 amino acids.
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Endotoxin Content:≤1 EU/µg protein (LAL test)
- Protein Synonyms:MIP-2alpha|CXCL2|GRO2|MGSAbeta
- Protein/Peptide Name:GRO-beta
- Purity:>95% (SDS-PAGE)
- Molecular Weight:~7.9 kDa
- Formulation:Lyophilized from a sterile (0.2 micron) filtered solution containing 0.1% Trifluoroacetic Acid (TFA).