Human Recombinant FGF-10 (from E. coli)
Supplier: Enzo Life Sciences
Certificates
About this item
Produced in E. coli. Contains 170 amino acids.
Specifications
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Endotoxin Content:≤1 EU/µg protein (LAL test)
- Biological Activity:4MBr-5 cell proliferation; ED50 ≤ 200 ng/ml (≥5.0×10E3 units/mg).
- Protein Synonyms:Fibroblast growth factor 10|Keratinocyte growth factor 2
- Protein/Peptide Name:FGF-10
- Purity:>95% (SDS-PAGE)
- Molecular Weight:~19.3 kDa
- Formulation:Lyophilized from a sterile (0.2 micron) filtered solution containing 10 mM sodium phosphate, pH 7.5