About this item
The Wizard Magnetic DNA Purification System for Food is designed for purification of DNA from a variety of food samples.
- Rapid Isolation of DNA from Many Food Types
- Requires minimal centrifugation and eliminates organic extractions
- Purified DNA suitable for PCR-based GMO testing
The Wizard Magnetic DNA Purification System for Food is designed for purification of DNA from a variety of food samples including corn seeds, cornmeal, soybeans, soy flour and soy milk. Processed food, such as corn chips, chocolate and chocolate-containing foods, lecithin and vegetable oils may also be used with the suggested protocol variations. The DNA purified from these samples can be used in PCR-based testing for genetically modified organism (GMO) DNA sequences.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: