About this item
ViviRen Live Cell Substrate is a uniquely engineered coelenterazine that generates three- to fivefold brighter Renilla luciferase luminescence for super-sensitive quantitation of reporter gene, BRET and RNAi activity.
- Sensitive Quantitation of Renilla Luminescence in Live Cells
- Engineered coelenterazine for live-cell reporter gene, BRET and RNAi assays
- Permits real-time measurement in multiwell plates
- High signal-to-background ratios
ViviRen Live Cell Substrate is a uniquely engineered coelenterazine that generates three- to fivefold brighter Renilla luciferase luminescence than wildtype coelenterazine. Using live cells, achieve up to 100-fold higher signal-to-noise ratios for super-sensitive quantitation of reporter gene, BRET and RNAi activity.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Temperature:LESS THAN -65°C