Specifications
- Description:Vac-Man Jr. Laboratory Vacuum Manifold, 2-sample capacity, 1 each
- Size:1 each
- Cat. No.:PAA7660
- Fits:Any column-based Wizard® purification system
- Supplier no.:A7660
Specifications
About this item
The ideal system for rapid, effective nucleic acid purification. Each manifold comes with 2 individually controlled One-Way Luer-Lok Stopcocks; up to 2 samples can be processed at a time.
Simultaneously process up to 2 nucleic acid samples
Use with standard 1- to 2-liter sidearm flasks
Eliminate need to handle waste during nucleic acid purification