DNA (Deoxyribonucleic acid) lambda, unmethylated
Catalog # PAD1521
Supplier: Promega Corporation
CAS Number:
Specifications
- Size:250 μg
- Cat. No.:PAD1521
- Supplier no.:D1521
Specifications
About this item
:
Storage Temperature:Freezer
CAS Number:9007-49-2
:73049-39-5
MDL Number:MFCD00130922