Order Entry
Puerto Rico
ContactUsLinkComponent
Transcend Colorimetric Translation Detection System, 30 Reactions, Promega
Transcend Colorimetric Translation Detection System, 30 Reactions, Promega
Catalog # PAL5070
CAS Number:  
Transcend Colorimetric Translation Detection System, 30 Reactions, Promega
Catalog # PAL5070
Supplier Number:  L5070
CAS Number:  

Specifications

  • Description:
    Transcend Colorimetric Translation Detection System, 30 reactions
  • Size:
    30 reactions
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAL5070
  • Supplier no.:
    L5070

Specifications

About this item

Non-Radioactive Translation Detection Systems for colorimetric or chemiluminescent detection of proteins synthesized in vitro. Biotinylated proteins are detected using Streptavidin-Alkaline Phosphatase or Streptavidin-HRP.

  • Non-Radioactive Detection of In Vitro Translated Proteins
  • Biotin tag allows detection of 0.5-5ng of protein 3-4 hours after gel electrophoresis
  • No radioisotope handling, storage or disposal issues
  • Option of colorimetric or chemiluminescent detection

The Transcend Non-Radioactive Translation Detection Systems allow non-radioactive detection of proteins synthesized in vitro. Using these systems, biotinylated lysine residues are incorporated into nascent proteins during translation, eliminating the need for labeling with [35S]methionine or other radioactive amino acids. This biotinylated lysine is added to the translation reaction as a precharged epsilon-labeled biotinylated lysine-tRNA complex (Transcend tRNA) rather than a free amino acid. After SDS-PAGE and electroblotting, the biotinylated proteins can be visualized by binding either Streptavidin-Alkaline Phosphatase (Streptavidin-AP) or Streptavidin-Horseradish Peroxidase (Streptavidin-HRP), followed either by colorimetric or chemiluminescent detection. Typically, these methods can detect 0.5-5ng of protein within 3-4 hours after gel electrophoresis. This sensitivity is equivalent to that achieved with [35S]methionine incorporation and autoradiographic detection 6-12 hours after gel electrophoresis.