Order Entry
Puerto Rico
ContactUsLinkComponent
TNT T7/SP6 Coupled Wheat Germ Extract System, 40 reactions, Promega
TNT T7/SP6 Coupled Wheat Germ Extract System, 40 reactions, Promega
Catalog # PAL5030
CAS Number:  
undefined
TNT T7/SP6 Coupled Wheat Germ Extract System, 40 reactions, Promega
Catalog # PAL5030
Supplier Number:  L5030
CAS Number:  

Specifications

  • Description:
    TNT T7/SP6 Coupled Wheat Germ Extract System, 40 reactions
  • Size:
    40 reactions
  • Storage Temperature:
    LESS THAN -65°C
  • Cat. No.:
    PAL5030
  • Supplier no.:
    L5030

Specifications

About this item

The TNT Coupled Wheat Germ Extract Systems are one-tube, coupled transcription/translation systems for eukaryotic cell-free protein expression.

  • Single-Tube Coupled Transcription/Translation System
  • Produces 2- to 6-fold more protein than standard wheat germ systems
  • Produce protein from linearized DNA in 1.5 hours
  • You choose the promoter

The TNT Coupled Wheat Germ Extract Systems offer researchers an alternative for eukaryotic cell-free protein expression: a one-tube, coupled transcription/translation system. The TNT Extract Systems greatly simplify the process and reduce the time required to obtain in vitro translation results. Standard wheat germ extract translations commonly use RNA synthesized in vitro from SP6 or T7 RNA polymerase promoters. This entire process requires separate reactions with several steps between each reaction. The TNT Extracts bypass many of these steps by incorporating transcription directly in the translation mix. Additionally, the TNT Extract reactions often produce significantly more protein (two- to sixfold) in a 1.5-hour reaction than do standard in vitro wheat germ extract translations using RNA templates. Magnesium Acetate, 25mM, and Potassium Chloride, 2.5M, can be used to optimize in vitro translation reactions in the TNT T7 Quick Coupled Transcription/Translation System, Flexi Rabbit Reticulocyte Lysate System and TNT Coupled Wheat Germ Extract System.