Order Entry
Puerto Rico
ContactUsLinkComponent
TNT T7 Quick Starter Bundle, Colorimetric, 1 each, Promega
TNT T7 Quick Starter Bundle, Colorimetric, 1 each, Promega
Catalog # PAL1215
CAS Number:  
TNT T7 Quick Starter Bundle, Colorimetric, 1 each, Promega
Catalog # PAL1215
Supplier Number:  L1215
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    TNT T7 Quick Starter Bundle, Colorimetric, 1 each
  • Size:
    1 each
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAL1215
  • Supplier no.:
    L1215

Specifications

About this item

Purchase this bundle, and get the TNT T7 Quick Coupled Transcription/Translation System, your choice of Transcend Translation Detection System, and receive two cell-free expression-qualified expression vectors, pTNT and pCMVTNT Vectors, at no extra cost.

  • Starter Pack for Protein Expression, Detection and Interactions Analysis
  • TnT(R) T7 Quick Coupled Transcription/Translation System, a simple one-hour, one-tube reaction
  • Transcend™ Translation Detection Systems detects proteins with either chemiluminescent or colorimetric techniques
  • pTnT™ and pCMVTnT™ Vectors work to enhance cell-free expression

Get the tools you need in one bundle to start cell-free expression and detection of your proteins of interest. Use the system for in vitro analysis of protein:protein or protein:nucleic acid interactions, or simply verify the ability of your clone to express protein. Purchase this special bundle, and get the popular TNT T7 Quick Coupled Transcription/Translation System, your choice of Transcend Translation Detection System, and receive two cell-free expression-qualified expression vectors, pTNT and pCMVTNT Vectors, at no extra cost.