- Description:TNT T7 Quick for PCR DNA, 40 reactions
- Size:40 reactions
- Storage Temperature:LESS THAN -65°C
- Cat. No.:PAL5540
- Supplier no.:L5540
TNT T7 Quick for PCR DNA is a rapid, convenient, coupled transcription/translation system designed for optimum protein expression from PCR templates.
- Coupled Transcription/Translation System for Optimum Protein Expression from PCR Templates
- Express protein directly from PCR, no cleanup needed
- Up to 5 times more protein than standard translation reactions with linear templates
- Includes Recombinant RNasin(R) Ribonuclease Inhibitor
TNT T7 Quick for PCR DNA is a rapid, convenient, coupled transcription/translation system designed for optimum protein expression from PCR templates. For most PCR templates, the TNT T7 Quick for PCR DNA reactions produce up to 5 times more protein than other commercially available kits. The PCR-generated DNA can be used directly from the amplification reaction or purified by numerous commercially available kits and traditional methods.