About this item
TGF beta R1 is a member of the TGF beta receptor subfamily and is a ser/thr protein kinase that forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF beta signal from the cell surface to the cytoplasm.
- Easily Screen and Profile TGFβR1 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant human TGF beta R1 (ALK5) (80-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TGF beta R1 or transforming growth factor beta-receptor 1 is a member of the TGF beta receptor subfamily and is a ser/thr protein kinase that forms a heteromeric complex with type II TGF-beta receptors when bound to TGF-beta, transducing the TGF beta signal from the cell surface to the cytoplasm. Mutations in the TGF beta R1 gene have been associated with Marfan syndrome, Loeys-Deitz Aortic Aneurysm Syndrome and the development of various types of tumors (1). TGF beta R1-dependent signaling is required for angiogenesis but not for the development of hematopoietic progenitor cells and functional hematopoiesis (2). ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: TGF beta R1, 10ug (Human, recombinant; amino acids 80-end). MW: ~66kDa. Substrate: TGFBR1 Peptide (KKKVLTQMGSPSIRC-S(pS)VS); derived from human SMAD3 (215-230). Other: Reaction Buffer, DTT. TGF beta R1 NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/7046/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Temperature:LESS THAN -65°C