- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:BloodCultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Description:SV Total RNA Isolation Start-Up Kit, 110V Electrical (250 preps, manifold and free vacuum pump), 1 each
- Size:1 each
- Storage Conditions:15°C to 30°C
- Description:SV Total RNA Isolation Start-Up Kit
- Cat. No.:PAZ3380
- Supplier no.:Z3380
A fast, simple technique for preparation of intact total RNA from tissues, cultured cells and white blood cells in as little as one hour.
- Bundle of 250 RNA Preps, Vacuum Manifold and Free Vacuum Pump
- Fast, simple method to extract total RNA from tissues, cultured cells and white blood cells
- Rapid isolation of high yields of total RNA without the use of hazardous compounds like phenol
- Two methods available for purification: microcentrifugation (spin) or vacuum
The SV Total RNA Isolation System provides a fast and simple technique for preparation of intact total RNA from tissues, cultured cells and white blood cells in as little as one hour. Using this membrane-based purification system, up to 60mg of tissue can be processed per purification, depending on tissue type. The system incorporates a DNase treatment step directly on the minicolumn membrane. This step substantially reduces genomic DNA contamination, which can interfere with amplification-based methodologies. Purification is achieved without the use of phenol:chloroform extractions or ethanol precipitations, and there is no DNase carryover in the final RNA preparation.