Order Entry
Puerto Rico
ContactUsLinkComponent
Single-Use HB101 Competent Cells, >10(8) cfu/ug, 1ml, Promega
Single-Use HB101 Competent Cells, >10(8) cfu/ug, 1ml, Promega
Catalog # PAL2015
CAS Number:  
undefined
Single-Use HB101 Competent Cells, >10(8) cfu/ug, 1ml, Promega
Catalog # PAL2015
Supplier Number:  L2015
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Single-Use HB101 Competent Cells, >10(8)cfu/ug, 1ml
  • Size:
    1ml
  • Storage Conditions:
    LESS THAN -65°C
  • Cat. No.:
    PAL2015
  • Supplier no.:
    L2015

Specifications

About this item

HB101 cells are recA- to minimize recombination events and contain a restriction-minus background (hsdS20) to allow better representation when cloning methylated DNA. Suitable for vectors that do not require alpha-complementation for blue/white screening.

  • Ready-to-Use Competent E. coli Cells for Recombinant Cloning and Subcloning
  • Cells are ready to use—no preparation time necessary
  • Transformation efficiencies guaranteed
  • No worries about undesirable recombination events (recA- mutation)

HB101 cells are recA-, which minimizes recombination events, and contain a restriction-minus background (hsdS20) that allows better representation when cloning methylated DNA. HB101 Competent Cells are suitable for vectors that do not require alpha-complementation for blue/white screening. For researchers doing more than one transformation, competent cells are available in standard format (200ul aliquots). For added convenience, single-use competent cells (50ul aliquots) also are offered. HB101 Competent Cells are prepared according to a modified procedure of Hanahan. HB101 Genotype: F-, thi-1, hsdS20 (r(B)-, m(B)-), supE44, recA13, ara-14, leuB6, proA2, lacY1, galK2, rpsL20 (str(r)), xyl-5, mtl-1.