Order Entry
Puerto Rico
ContactUsLinkComponent
Single-Use BL21(DE3)pLysS Competent Cells, 1 ml, Promega
Single-Use BL21(DE3)pLysS Competent Cells, 1 ml, Promega
Catalog # PAL1195
CAS Number:  
undefined
Single-Use BL21(DE3)pLysS Competent Cells, 1 ml, Promega
Catalog # PAL1195
Supplier Number:  L1195
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Single-Use BL21(DE3)pLysS Competent Cells, 1ml
  • Size:
    1ml
  • Storage Conditions:
    Store at –70°C
  • Cat. No.:
    PAL1195
  • Supplier no.:
    L1195

Specifications

About this item

BL21(DE3)pLysS Competent Cells allow high-efficiency protein expression of any gene that is under the control of a T7 promoter and has a ribosome binding site.

  • High-Efficiency Protein Expression from Gene with T7 Promoter and Ribosome Binding Site
  • Inducible protein expression from gene with T7 RNA polymerase controlled by lac UV5 promoter
  • Increased stability of expressed protein from deficiency in proteases lon and OmpT
  • Lower background expression of target genes with pLysS plasmid

BL21(DE3)pLysS Competent Cells allow high-efficiency protein expression of any gene that is under the control of a T7 promoter and has a ribosome binding site. BL21(DE3)pLysS is lysogenic for lambda-DE3, which contains the T7 bacteriophage gene I, encoding T7 RNA polymerase under the control of the lac UV5 promoter. BL21(DE3)pLysS also contains a plasmid, pLysS, which carries the gene encoding T7 lysozyme. T7 lysozyme lowers the background expression level of target genes under the control of the T7 promoter but does not interfere with the level of expression achieved following induction by IPTG. For researchers doing more than one transformation, competent cells are available in standard format (200ul aliquots). For added convenience, single-use competent cells (50ul aliquots) also are offered. Genotype: F-, ompT, hsdS(B) (r(B)-, m(B)-), dcm, gal, lambda(DE3), pLysS, Cm(r).