Riboprobe System-T3, 1 system, Promega
Catalog # PAP1430
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:Riboprobe System-T3, 1 system
- Size:1 system
- Cat. No.:PAP1430
- Supplier no.:P1430
Specifications
About this item
The Riboprobe Systems are designed to prepare high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts.
- Produce High-Specific-Activity RNA Probes or Defined RNA Transcripts
- Systems available with SP6 RNA Polymerase, T7 RNA Polymerase or T3 RNA Polymerase
- SP6, T7 and T3 RNA Polymerases are extremely promoter-specific, allowing production of virtually homogeneous RNA using plasmid DNA as a template
- Allow synthesis of RNA corresponding to the coding or noncoding strand of cloned DNA from a single plasmid construct
The Riboprobe Systems are designed for in vitro preparation of high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts. These systems contain all components necessary for in vitro transcription from a DNA template (excluding the radioisotope) and also contain RQ1 RNase-Free DNase (Cat.# M6101) for template removal following transcription.
Caution: Store at –20°C