Riboprobe Combination System-T3/T7 RNA Polymerase, 1 system, Promega
PAP1450
:
- Description:Riboprobe Combination System-T3/T7 RNA Polymerase, 1 system
- Size:1 system
- Storage Conditions:-30°C to -10°C
- Cat. No.:PAP1450
- Supplier no.:P1450
The Riboprobe Systems are designed to prepare high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts.
- Produce High-Specific-Activity RNA Probes or Defined RNA Transcripts
- Choose from combination systems with T3/T7 or SP6/T7 RNA polymerases
- SP6, T7 and T3 RNA Polymerases are extremely promoter-specific, allowing production of virtually homogeneous RNA using plasmid DNA as a template
- Allows synthesis of RNA corresponding to the coding or noncoding strand of cloned DNA from a single plasmid construct
The Riboprobe Combination Systems are designed for in vitro preparation of high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts. The Riboprobe Combination Systems include the RNA polymerases, all of the required reagents (excluding radioisotope) for performing transcription reactions in vitro and RQ1 RNase-Free DNase (Cat.# M6101) for removal of the template following transcription.